Butylamine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L03575-0F
L03575-AE
L03575-AP
CAAAAL03575-0FEA
135.56
CAD
CAAAAL03575-0F
CAAAAL03575-AE
CAAAAL03575-AP
Butylamine 99%
Butylamine
Purity: 99%
CAS No.: 109-73-9
Molecular Formula: C4H11N
Formula Weight: 73.14
Flash Point: -14°(6°F)
Refractive Index: 1.4010
Odor: Ammonia-like
Melting Point: -50°
Boiling Point: 76-78°
Beilstein Reference: 605269
Merck Reference: 14,1543
UN No.: 1125
MDL No.: MFCD00011690
Formula:
H₃C(CH₂)₂CH₂NH₂ Boiling Pt: 76…78 °C Melting Pt: –50 °C Density: 0.738 Flash Pt: –14 °C (6 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011690 CAS Number: 109-73-9 EINECS: 203-699-2 UN: 1125 ADR: 3,II Merck Index: 14,1543 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...