2,4-Dimethylpentane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L03570-04
L03570-09
CAAAAL03570-04EA
97.49
CAD
CAAAAL03570-04
CAAAAL03570-09
2,4-Dimethylpentane 98+%
2,4-Dimethylpentane
Purity: 98+%
CAS No.: 108-08-7
Molecular Formula: C7H16
Formula Weight: 100.21
Flash Point: -12°(10°F)
Refractive Index: 1.3820
Melting Point: ca -123°
Boiling Point: 79-80°
Beilstein Reference: 1696855
UN No.: 1206
MDL No.: MFCD00008945
Formula:
C₇H₁₆ Boiling Pt: 79…80 °C Melting Pt: ∼ 123 °C Density: 0.670 Flash Pt: –12 °C (10 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008945 CAS Number: 108-08-7 EINECS: 203-548-0 UN: 1206 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...