(2-Cyanoethyl)triethoxysilane 97%
Supplier: Thermo Scientific Chemicals
L03543-14
L03543-22
CAAAAL03543-14EA
67.47
CAD
CAAAAL03543-14
CAAAAL03543-22
(2-Cyanoethyl)triethoxysilane 97%
(2-Cyanoethyl)triethoxysilane
Purity: 97%
CAS No.: 919-31-3
Molecular Formula: NCCH2CH2Si(OCH2CH3)3
Formula Weight: 217.34
Flash Point: 100°(212°F)
Refractive Index: 1.4130
Boiling Point: 224-225°
Beilstein Reference: 1776392
MDL No.: MFCD00019851
Formula:
NCCH₂CH₂Si(OCH₂CH₃)₃ Boiling Pt: 224…225 °C Density: 0.980 Flash Pt: 100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00019851 CAS Number: 919-31-3 EINECS: 213-050-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...