Octanoyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

L03526-AE L03526-AP
CAAAAL03526-AEEA 50.09 CAD
CAAAAL03526-AE CAAAAL03526-AP
Octanoyl chloride 99%
Octanoyl chloride

Purity: 99% CAS No.: 111-64-8 Molecular Formula: CH3(CH2)6COCl Formula Weight: 162.66 Flash Point: 80°(176°F) Refractive Index: 1.4350 Odor: Pungent Melting Point: -63° Boiling Point: 195° Beilstein Reference: 635917 UN No.: 3265 MDL No.: MFCD00000766

Formula: C₈H₁₅ClO
MW: 162.66 g/mol
Boiling Pt: 195 °C
Melting Pt: –63 °C
Density: 0.942
Flash Pt: 80 °C (176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000766
CAS Number: 111-64-8
EINECS: 203-891-6
UN: 2927
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR