N,N-Diethyl-1,1-dimethylsilylamine 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: DMSDEA, N,N-Diethyldimethylsilylamine, N-(Dimethylsilyl)diethylamine, N,N-Diethylaminodimethylsilane, Dimethylsilyldiethylamine, (Diethylamino)dimethylsilane

L03503-06 L03503-14
CAAAAL03503-06EA 94.48 CAD
CAAAAL03503-06 CAAAAL03503-14
N,N-Diethyl-1,1-dimethylsilylamine 95%
N,N-Diethyl-1,1-dimethylsilylamine

Purity: 95% CAS No.: 13686-66-3 Molecular Formula: C6H17NSi Formula Weight: 131.30 Flash Point: -5°(23°F) Refractive Index: 1.4080 Boiling Point: 109-110° Beilstein Reference: 741862 UN No.: 1993 MDL No.: MFCD00048523

Formula: C₆H₁₇NSi
MW: 131.29 g/mol
Boiling Pt: 109…110 °C
Density: 0.800
Flash Pt: <–5 °C (23 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048523
CAS Number: 13686-66-3
EINECS: 237-202-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR