Diethyl sulfide 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L03493-09
L03493-22
CAAAAL03493-09EA
71.89
CAD
CAAAAL03493-09
CAAAAL03493-22
Diethyl sulfide 96%
Diethyl sulphide
Purity: 96%
CAS No.: 352-93-2
Molecular Formula: CH3CH2SCH2CH3
Formula Weight: 90.19
Flash Point: -10°(14°F)
Refractive Index: 1.4420
Odor: Garlic-like
Melting Point: -104°
Boiling Point: 90-92°
Beilstein Reference: 1696909
Merck Reference: 14,3854
UN No.: 2375
MDL No.: MFCD00009270
Formula:
C₄H₁₀S MW: 90.19 g/mol Boiling Pt: 90…92 °C Melting Pt: –104 °C Density: 0.835 Flash Pt: –10 °C (14 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009270 CAS Number: 352-93-2 EINECS: 206-526-9 UN: 2375 ADR: 3 II Merck Index: 14,3854 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...