3,4-Lutidine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L03489-AC
L03489-AE
L03489-AP
CAAAAL03489-ACEA
49.93
CAD
CAAAAL03489-AC
CAAAAL03489-AE
CAAAAL03489-AP
3,4-Lutidine 98+%
3,4-Lutidine
Purity: 98+%
CAS No.: 583-58-4
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 47°(116°F)
Refractive Index: 1.5120
Odor: Unpleasant
Melting Point: -11°
Boiling Point: 163-164°
Beilstein Reference: 106583
UN No.: 2929
MDL No.: MFCD00006403
Formula:
C₇H₉N Boiling Pt: 163…164 °C Melting Pt: –11 °C Density: 0.955 Flash Pt: 47 °C (116 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006403 CAS Number: 583-58-4 EINECS: 209-511-5 UN: 2929 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...