Butyl acrylate 98+% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: n-Butyl acrylate, Acrylic acid butyl ester

Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) ≤50 ppm

L03476-0F L03476-AE L03476-AP
CAAAAL03476-0FEA 167.48 CAD
CAAAAL03476-0F CAAAAL03476-AE CAAAAL03476-AP
Butyl acrylate 98+% stabilized
Butyl acrylate

Purity: 98+% CAS No.: 141-32-2 Molecular Formula: C7H12O2 Formula Weight: 128.17 Flash Point: 39°(102°F) Refractive Index: 1.4180 Odor: Fruit-like Melting Point: -64° Boiling Point: 145-146° Beilstein Reference: 1749970 Merck Reference: 14,1539 UN No.: 2348 MDL No.: MFCD00009446

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 145…146 °C
Melting Pt: –64 °C
Density: 0.895
Flash Pt: 39 °C (102 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008809
CAS Number: 141-32-2
EINECS: 205-480-7
UN: 2348
ADR: 3,III
Merck Index: 14,1539

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR