2,4,6-Trichloro-1,3,5-triazine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cyanuric chloride, 2,4,6-Trichloro-s-triazine

L03442-0B L03442-18 L03442-30
CAAAAL03442-0BEA 199.08 CAD
CAAAAL03442-0B CAAAAL03442-18 CAAAAL03442-30
2,4,6-Trichloro-1,3,5-triazine 98%
2,4,6-Trichloro-1,3,5-triazine

Reagent for detection of glycine in presence of other amino acids


Purity: 98% CAS No.: 108-77-0 Molecular Formula: C3Cl3N3 Formula Weight: 184.41 Storage Temperature: 2°C to 8°C Flash Point: 190°(374°F) Melting Point: 145-149° Boiling Point: 192-194° Beilstein Reference: 124246 UN No.: 2670 MDL No.: MFCD00006046

Formula: C₃Cl₃N₃
MW: 184.41 g/mol
Boiling Pt: 190
Melting Pt: 145…149 °C
Density: 1.920
Flash Pt: >190 °C (374 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00006046
CAS Number: 108-77-0
EINECS: 203-614-9
UN: 2670
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR