Methylsulfonylacetone 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-(Methylsulphonyl)propanone, 1-(Methylsulfonyl)propanone
L03421-04
L03421-09
L03421-18
CAAAAL03421-04EA
46.93
CAD
CAAAAL03421-04
CAAAAL03421-09
CAAAAL03421-18
Methylsulfonylacetone 98+%
Methylsulphonylacetone
Purity: 98+%
CAS No.: 5000-46-4
Molecular Formula: C4H8O3S
Formula Weight: 136.18
Melting Point: 47-52°
Beilstein Reference: 1754113
Formula:
C₄H₈O₃S Melting Pt: 47…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014745 CAS Number: 5000-46-4 EINECS: 225-665-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...