3,5-Dichloropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03380-14
L03380-22
CAAAAL03380-14EA
69.2
CAD
CAAAAL03380-14
CAAAAL03380-22
3,5-Dichloropyridine 98%
3,5-Dichloropyridine
Purity: 98%
CAS No.: 2457-47-8
Molecular Formula: C5H3Cl2N
Formula Weight: 147.99
Flash Point: 74°(165°F)
Odor: Unpleasant
Boiling Point: 178°
Beilstein Reference: 1973
UN No.: 2811
MDL No.: MFCD00006376
Formula:
C₅H₃Cl₂N Boiling Pt: 178 °C Melting Pt: 63…67 °C Density: 1.39 Flash Pt: 74 °C(165 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006376 CAS Number: 2457-47-8 EINECS: 219-537-9 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...