Allylthiourea 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-Allylthiourea, Tiosinamine, 1-Allyl-2-thiourea, Allyl thiourea, Rhodalline, Thiosinamine, N-Allylthiourea
L03377-14
L03377-22
CAAAAL03377-14EA
60.36
CAD
CAAAAL03377-14
CAAAAL03377-22
Allylthiourea 98%
Allylthiourea
Purity: 98%
CAS No.: 109-57-9
Molecular Formula: C4H8N2S
Formula Weight: 116.19
Odor: Unpleasant
Melting Point: 70-78°
Beilstein Reference: 1071470
Merck Reference: 14,9362
UN No.: 2811
MDL No.: MFCD00004940
Formula:
H₂CCHCH₂NHC(S)NH₂ Boiling Pt: 191 °C (1013 hPa) Melting Pt: 70…78 °C Density: 1.220 Storage Temperature: Ambient |
MDL Number:
MFCD00004940 CAS Number: 109-57-9 EINECS: 203-683-5 UN: 2811 ADR: 6.1,III Merck Index: 14,9362 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...