α-Bromo-p-toluic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromomethylbenzoic acid, 4-(Bromomethyl)benzoic acid

L03371-06 L03371-14 L03371-22
CAAAAL03371-06EA 71.42 CAD
CAAAAL03371-06 CAAAAL03371-14 CAAAAL03371-22
α-Bromo-p-toluic acid 97%
α-Bromo-p-toluic acid

Purity: 97% CAS No.: 6232-88-8 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Melting Point: 223-227° Beilstein Reference: 1862870 UN No.: 3261 MDL No.: MFCD00002567

Formula: C₈H₇BrO₂
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 223…227 °C
Storage Temperature: Ambient
MDL Number: MFCD00002567
CAS Number: 6232-88-8
EINECS: 228-343-3
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR