Allyl phenyl ether 99%
Supplier: Thermo Scientific Chemicals
Warning
|
L03359-14
L03359-22
CAAAAL03359-14EA
94.48
CAD
CAAAAL03359-14
CAAAAL03359-22
Allyl phenyl ether 99%
Allyl phenyl ether
Purity: 99%
CAS No.: 1746-13-0
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 62°(143°F)
Refractive Index: 1.5210
Odor: Characteristic
Boiling Point: 191-192°
Beilstein Reference: 1905622
MDL No.: MFCD00008644
Formula:
H₂CCHCH₂OC₆H₅ Boiling Pt: 191…192 °C Density: 0.979 Flash Pt: 62 °C(143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008644 CAS Number: 1746-13-0 EINECS: 217-125-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...