4-Aminophthalic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

L03333-03 L03333-06
CAAAAL03333-03EA 77.89 CAD
CAAAAL03333-03 CAAAAL03333-06
4-Aminophthalic acid 98%
4-Aminophthalic acid

Purity: 98% CAS No.: 5434-21-9 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Odor: Odorless Melting Point: 300° Beilstein Reference: 2723405 MDL No.: MFCD00013985

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00013985
CAS Number: 5434-21-9
EINECS: 226-596-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR