6-Aminouracil 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Amino-2,4-dihydroxypyrimidine, 6-Aminopyrimidine-2,4(1H,3H)-dione, 4-Amino-2,6-dihydroxypyrimidine, 4-Aminouracil

L03332-22 L03332-36
CAAAAL03332-22EA 97.17 CAD
CAAAAL03332-22 CAAAAL03332-36
6-Aminouracil 98%
6-Aminouracil

Purity: 98% CAS No.: 873-83-6 Molecular Formula: C4H5N3O2 Formula Weight: 127.10 Odor: Odorless Melting Point: 360° Beilstein Reference: 120491 MDL No.: MFCD00006071

Formula: C₄H₅N₃O₂
MW: 127.10 g/mol
Melting Pt: 360 °C
Storage Temperature: Ambient
MDL Number: MFCD00006071
CAS Number: 873-83-6
EINECS: 212-854-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR