2,4,5-Trichlorothiophenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03324-03
L03324-06
CAAAAL03324-03EA
74.26
CAD
CAAAAL03324-03
CAAAAL03324-06
2,4,5-Trichlorothiophenol 97%
2,4,5-Trichlorothiophenol
Purity: 97%
CAS No.: 3773-14-6
Molecular Formula: C6H3Cl3S
Formula Weight: 213.51
Odor: Unpleasant
Melting Point: 112-118°
Boiling Point: 153-154°/15mm
Beilstein Reference: 509727
UN No.: 3335
MDL No.: MFCD00014447
Formula:
C₆H₃Cl₃S Boiling Pt: 153…154 °C/15 mm Melting Pt: 112…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014447 CAS Number: 3773-14-6 EINECS: 223-223-7 UN: 3335 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...