4-Methoxychalcone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methoxybenzylidenacetophenone, 3-(4-Methoxyphenyl)-1-phenylprop-2-en-1-one

L03319-06 L03319-14
CAAAAL03319-06EA 40.13 CAD
CAAAAL03319-06 CAAAAL03319-14
4-Methoxychalcone 97%
4-Methoxychalcone

Purity: 97% CAS No.: 959-33-1 Molecular Formula: C16H14O2 Formula Weight: 238.29 Melting Point: 73-76° Beilstein Reference: 978742

Formula: C₁₆H₁₄O₂
Boiling Pt: 187 °C (19 mmHg)
Melting Pt: 73…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00017179
CAS Number: 959-33-1
EINECS: 213-499-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR