Heptanoyl chloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L03315-18
L03315-30
CAAAAL03315-18EA
71.89
CAD
CAAAAL03315-18
CAAAAL03315-30
Heptanoyl chloride 99%
Heptanoyl chloride
Purity: 99%
CAS No.: 2528-61-2
Molecular Formula: C7H13ClO
Formula Weight: 148.63
Flash Point: 58°(136°F)
Refractive Index: 1.4310
Odor: Acrid
Melting Point: -84°
Boiling Point: 172-174°
Beilstein Reference: 605636
UN No.: 2920
MDL No.: MFCD00000763
Formula:
C₇H₁₃ClO Boiling Pt: 172…174 °C Melting Pt: –84 °C Density: 0.960 Flash Pt: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000763 CAS Number: 2528-61-2 EINECS: 219-775-3 UN: 2927 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...