Thioacetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L03305-14
L03305-22
L03305-36
CAAAAL03305-14EA
53.4
CAD
CAAAAL03305-14
CAAAAL03305-22
CAAAAL03305-36
Thioacetic acid 97%
Thioacetic acid
Purity: 97%
CAS No.: 507-09-5
Molecular Formula: C2H4OS
Formula Weight: 76.12
Storage Temperature: 2°C to 8°C
Flash Point: 18°(64°F)
Refractive Index: 1.4640
Odor: Stench
Melting Point: -17°
Boiling Point: 97°
Beilstein Reference: 773684
Merck Reference: 14,9320
UN No.: 2436
MDL No.: MFCD00004853
Formula:
C₂H₄OS MW: 76.12 g/mol Boiling Pt: 97 °C Melting Pt: –17 °C Density: 1.068 Flash Pt: 18 °C (64 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004853 CAS Number: 507-09-5 EINECS: 208-063-8 UN: 2436 ADR: 3,II Merck Index: 14,9320 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...