Benzyl alcohol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03292-0B
L03292-0E
L03292-30
CAAAAL03292-0BEA
128.61
CAD
CAAAAL03292-0B
CAAAAL03292-0E
CAAAAL03292-30
Benzyl alcohol 99%
Benzyl alcohol
Purity: 99%
CAS No.: 100-51-6
Molecular Formula: C7H8O
Formula Weight: 108.14
Flash Point: 100°(212°F)
Refractive Index: 1.5400
Odor: Aromatic
Melting Point: -15°
Boiling Point: 205°
Beilstein Reference: 878307
Merck Reference: 14,1124
MDL No.: MFCD00004599
Formula:
C₆H₅CH₂OH Boiling Pt: 205 °C Melting Pt: –15 °C Density: 1.045 Flash Pt: 100 °C(212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004599 CAS Number: 100-51-6 EINECS: 202-859-9 Merck Index: 14,1124 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...