8-Methylquinoline 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03288-06
L03288-14
CAAAAL03288-06EA
75.37
CAD
CAAAAL03288-06
CAAAAL03288-14
8-Methylquinoline 97+%
8-Methylquinoline
Purity: 97+%
CAS No.: 611-32-5
Molecular Formula: C10H9N
Formula Weight: 143.19
Flash Point: 105°(221°F)
Refractive Index: 1.6150
Odor: Unpleasant
Melting Point: -80°
Boiling Point: 134-136°/25mm
Beilstein Reference: 111340
MDL No.: MFCD00006810
Formula:
C₁₀H₉N MW: 143.19 g/mol Boiling Pt: 134…136 °C (25 mmHg) Melting Pt: –80 °C Density: 1.074 Flash Pt: 105 °C (221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006810 CAS Number: 611-32-5 EINECS: 210-264-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...