Benzyl benzoate 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Benzoic acid benzyl ester
L03258-0B
L03258-0E
L03258-30
CAAAAL03258-0BEA
77.1
CAD
CAAAAL03258-0B
CAAAAL03258-0E
CAAAAL03258-30
Benzyl benzoate 99+%
Benzyl benzoate
Purity: 99+%
CAS No.: 120-51-4
Molecular Formula: C14H12O2
Formula Weight: 212.25
Flash Point: 147°(296°F)
Refractive Index: 1.5680
Melting Point: 18-20°
Boiling Point: 323-324°
Beilstein Reference: 2049280
Merck Reference: 14,1127
UN No.: 3082
MDL No.: MFCD00003075
Formula:
C₁₄H₁₂O₂ MW: 212.25 g/mol Boiling Pt: 323 Melting Pt: 21 Density: 1.112 Flash Pt: 147 °C (296 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003075 CAS Number: 120-51-4 EINECS: 204-402-9 UN: 3082 ADR: 9 III Merck Index: 14,1127 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...