Adonitol 99%
Supplier: Thermo Scientific Chemicals
L03253-06
L03253-14
CAAAAL03253-06EA
50.24
CAD
CAAAAL03253-06
CAAAAL03253-14
Adonitol 99%
Adonitol
Purity: 99%
CAS No.: 488-81-3
Molecular Formula: C5H12O5
Formula Weight: 152.15
Melting Point: 102-104°
Beilstein Reference: 1720524
Merck Reference: 14,168
MDL No.: MFCD00064291
Formula:
HOH₂C(CH(OH))₃CH₂OH Melting Pt: 102…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064291 CAS Number: 488-81-3 EINECS: 207-685-7 Merck Index: 14,168 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...