Ethyl nicotinate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03215-22
L03215-36
CAAAAL03215-22EA
69.05
CAD
CAAAAL03215-22
CAAAAL03215-36
Ethyl nicotinate 99%
Ethyl nicotinate
Purity: 99%
CAS No.: 614-18-6
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Flash Point: 93°(199°F)
Refractive Index: 1.5030
Odor: Characteristic
Melting Point: 8-9°
Boiling Point: 223-224°
Beilstein Reference: 122937
MDL No.: MFCD00006389
Formula:
C₈H₉NO₂ Boiling Pt: 223…224 °C Melting Pt: 8…9 °C Density: 1,107 Flash Pt: 93 °C(199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006389 CAS Number: 614-18-6 EINECS: 210-370-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...