(±)-2-Methyl-2,4-pentanediol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03187-0F
L03187-AE
L03187-AP
CAAAAL03187-0FEA
112.02
CAD
CAAAAL03187-0F
CAAAAL03187-AE
CAAAAL03187-AP
(±)-2-Methyl-2,4-pentanediol 98%
(±)-2-Methyl-2,4-pentanediol
Purity: 98%
CAS No.: 107-41-5
Molecular Formula: C6H14O2
Formula Weight: 118.18
Flash Point: 93°(199°F)
Refractive Index: 1.4270
Odor: Weak
Melting Point: -40°
Boiling Point: 197°
Beilstein Reference: 1098298
Merck Reference: 14,4710
MDL No.: MFCD00004547
Formula:
C₆H₁₄O₂ Boiling Pt: 197 °C Melting Pt: –40 °C Density: 0.922 Flash Pt: 93 °C(199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004547 CAS Number: 107-41-5 EINECS: 203-489-0 Merck Index: 14,4710 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...