4-Nitro-m-cresol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03163-14
L03163-22
CAAAAL03163-14EA
69.36
CAD
CAAAAL03163-14
CAAAAL03163-22
4-Nitro-m-cresol 98%
4-Nitro-m-cresol
Purity: 98%
CAS No.: 2581-34-2
Molecular Formula: C7H7NO3
Formula Weight: 153.14
Odor: Phenol-like
Melting Point: 126-130°
Beilstein Reference: 1868105
MDL No.: MFCD00007333
Formula:
C₇H₇NO₃ Melting Pt: 126…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007333 CAS Number: 2581-34-2 EINECS: 219-952-5 UN: 2446 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...