Tritylium hexafluorophosphate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Triphenylcarbenium hexafluorophosphate
L03140-09
L03140-18
CAAAAL03140-09EA
222.78
CAD
CAAAAL03140-09
CAAAAL03140-18
Tritylium hexafluorophosphate 98%
Tritylium hexafluorophosphate
Purity: 98%
CAS No.: 437-17-2
Molecular Formula: C19H15F6P
Formula Weight: 388.29
Storage Temperature: -30°C to -10°C
Melting Point: ca 160° dec.
Beilstein Reference: 4344297
UN No.: 3261
MDL No.: MFCD00013121
Formula:
C₁₉H₁₅F₆P MW: 388.29 g/mol Melting Pt: ∼ 160 °C (dec.) Storage Temperature: Freezer |
MDL Number:
MFCD00013121 CAS Number: 437-17-2 EINECS: 207-112-0 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...