5-Amino-2-naphthalenesulfonic acid 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 5-Amino-2-naphthalenesulphonic Acid, 5-Aminonaphthalene-2-sulfonic acid, 5-Amino-β-naphthalenesulfonic acid, 5-Amino-β-naphthalenesulphonic acid, 1-Naphthylamine-6-sulfonic acid, 1,6-Cleve's acid, 5-Amino-2-naphthalenesulfonic Acid, 5-Aminonaphthalene-2-sulphonic acid, 5-Aminonaphthalene-2-sulfonic acid

L03099-06 L03099-14
CAAAAL03099-06EA 43.13 CAD
CAAAAL03099-06 CAAAAL03099-14
5-Amino-2-naphthalenesulfonic acid 97%
5-Amino-2-naphthalenesulphonic acid

Purity: 97% CAS No.: 119-79-9 Molecular Formula: C10H9NO3S Formula Weight: 223.25 Odor: Odorless Melting Point: 300° Beilstein Reference: 1819887 Merck Reference: 14,2350 MDL No.: MFCD00004030

Formula: C₁₀H₉NO₃S
MW: 223.25 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004030
CAS Number: 119-79-9
EINECS: 204-351-2
Merck Index: 14,2350

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR