4-Bromo-o-toluic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Bromo-2-methylbenzoic acid

L03071-06 L03071-14
CAAAAL03071-03EA 43.48 CAD
CAAAAL03071-03 CAAAAL03071-06 CAAAAL03071-14
4-Bromo-o-toluic acid 98+%
4-Bromo-o-toluic acid

Purity: 98+% CAS No.: 68837-59-2 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Melting Point: 182-184° Beilstein Reference: 2249816

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Melting Pt: 182…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00040905
CAS Number: 68837-59-2
EINECS: 272-437-7
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR