1,2-Diiodoethane 98%

Supplier: Thermo Scientific Chemicals
Warning

L03019-06 L03019-14
CAAAAL03019-06EA 44.24 CAD
CAAAAL03019-06 CAAAAL03019-14
1,2-Diiodoethane 98%
1,2-Diiodoethane

Purity: 98% CAS No.: 624-73-7 Molecular Formula: ICH2CH2I Formula Weight: 281.86 Storage Temperature: 2°C to 8°C Melting Point: 80-84° Beilstein Reference: 1730870 MDL No.: MFCD00001093

Formula: C₂H₄I₂
Boiling Pt: 206 °C (1013 hPa)
Melting Pt: 80…84 °C
Density: 2,130
Storage Temperature: Refrigerator
MDL Number: MFCD00001093
CAS Number: 624-73-7
EINECS: 210-859-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR