Ethyl isonicotinate 98%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
4-Picolinic acid ethyl ester, 4-Pyridinecarboxylic acid ethyl ester, Pyridine-4-carboxylic acid ethyl ester, Ethyl 4-pyridinecarboxylate, Isonicotinic acid ethyl ester
L03010-22
L03010-36
CAAAAL03010-22EA
102.86
CAD
CAAAAL03010-22
CAAAAL03010-36
Ethyl isonicotinate 98%
Ethyl isonicotinate
Purity: 98%
CAS No.: 1570-45-2
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Flash Point: 87°(188°F)
Refractive Index: 1.5010
Odor: Characteristic
Boiling Point: 220°
Beilstein Reference: 122942
Merck Reference: 14,5187
MDL No.: MFCD00006428
Formula:
C₈H₉NO₂ MW: 151.16 g/mol Boiling Pt: 220 Density: 1.008 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006428 CAS Number: 1570-45-2 EINECS: 216-379-2 Merck Index: 14,5187 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...