Butyronitrile 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L02999-AK
L02999-AU
CAAAAL02999-AKEA
52.77
CAD
CAAAAL02999-AK
CAAAAL02999-AU
Butyronitrile 99%
Butyronitrile
Purity: 99%
CAS No.: 109-74-0
Molecular Formula: C4H7N
Formula Weight: 69.11
Flash Point: 18°(64°F)
Refractive Index: 1.3840
Melting Point: -112°
Boiling Point: 115-117°
Beilstein Reference: 1361452
Merck Reference: 14,1597
UN No.: 2411
MDL No.: MFCD00001968
Formula:
C₄H₇N Boiling Pt: 115…117 °C Melting Pt: –112 °C Density: 0.792 Flash Pt: 18 °C (64 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001968 CAS Number: 109-74-0 EINECS: 203-700-6 UN: 2411 ADR: 3,II Merck Index: 14,1597 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...