Methyl oleate, Technical Grade C18 71-90%, C18:1 >65% of C18
Supplier: Thermo Scientific Chemicals
L02937-AK
L02937-AU
CAAAAL02937-AKEA
41.87
CAD
CAAAAL02937-AK
CAAAAL02937-AU
Methyl oleate, Technical Grade C18 71-90%, C18:1 >65% of C18
Methyl oleate
CAS No.: 112-62-9
Molecular Formula: C19H36O2
Formula Weight: 296.50
Flash Point: 163°(325°F)
Refractive Index: 1.4520
Odor: Weak, characteristic
Boiling Point: 168-170°/2mm
Beilstein Reference: 1727037
Merck Reference: 14,6828
MDL No.: MFCD00009578
Formula:
C₁₉H₃₆O₂ Boiling Pt: 168…170 °C/2 mm Melting Pt: –20 °C Density: 0.876 Flash Pt: 163 °C(325 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009578 CAS Number: 112-62-9 EINECS: 203-992-5 Merck Index: 14,6828 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...