4'-Bromoacetanilide 98%

Supplier: Thermo Scientific Chemicals
Warning

L02924-18 L02924-30
CAAAAL02924-18EA 75.84 CAD
CAAAAL02924-18 CAAAAL02924-30
4'-Bromoacetanilide 98%
4'-Bromoacetanilide

Purity: 98% CAS No.: 103-88-8 Molecular Formula: C8H8BrNO Formula Weight: 214.06 Odor: Odorless Melting Point: 166-171° Beilstein Reference: 2208091 Merck Reference: 14,1399 MDL No.: MFCD00000092

Formula: H₃CC(O)NH(C₆H₄Br)
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 166…171 °C
Density: 1.717 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000092
CAS Number: 103-88-8
EINECS: 203-154-9
Merck Index: 14,1399

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR