3,4,5-Trimethylphenol 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L02911-03
L02911-06
L02911-14
CAAAAL02911-03EA
67.94
CAD
CAAAAL02911-03
CAAAAL02911-06
CAAAAL02911-14
3,4,5-Trimethylphenol 97%
3,4,5-Trimethylphenol
Purity: 97%
CAS No.: 527-54-8
Molecular Formula: C9H12O
Formula Weight: 136.20
Odor: Phenol-like
Melting Point: 104-108°
Beilstein Reference: 1907246
UN No.: 2430
MDL No.: MFCD00002305
Formula:
C₉H₁₂O Boiling Pt: 249 °C (1013 hPa) Melting Pt: 104…108 °C Density: 0.8184 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002305 CAS Number: 527-54-8 EINECS: 208-418-7 UN: 2430 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...