Allyl isothiocyanate 94% stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
L02901-06
L02901-18
L02901-30
CAAAAL02901-06EA
43.29
CAD
CAAAAL02901-06
CAAAAL02901-18
CAAAAL02901-30
Allyl isothiocyanate 94% stabilised
Allyl isothiocyanate
Purity: 94%
CAS No.: 57-06-7
Molecular Formula: CH2=CHCH2NCS
Formula Weight: 99.16
Storage Temperature: 2°C to 8°C
Flash Point: 46°(114°F)
Refractive Index: 1.5300
Odor: Pungent
Melting Point: -80°
Boiling Point: 149-151°
Beilstein Reference: 773748
Merck Reference: 14,295
UN No.: 1545
MDL No.: MFCD00004822
Formula:
C₄H₅NS MW: 99.16 g/mol Boiling Pt: 148 Melting Pt: -80 Density: 1.013 Flash Pt: 46 °C (114 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004822 CAS Number: 57-06-7 EINECS: 200-309-2 UN: 1545 ADR: 6.1 II Merck Index: 14,295 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...