2,4-Dimethylbenzyl alcohol 97%

Supplier: Thermo Scientific Chemicals

Synonyms: (2,4-Dimethylphenyl)methanol, 4-(Hydroxymethyl)-m-xylene

L02899-03 L02899-06
CAAAAL02899-03EA 63.67 CAD
CAAAAL02899-03 CAAAAL02899-06
2,4-Dimethylbenzyl alcohol 97%
2,4-Dimethylbenzyl alcohol

Purity: 97% CAS No.: 16308-92-2 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 110°(230°F) Refractive Index: 1.5360 Boiling Point: 120-121°/13mm Beilstein Reference: 1859679 MDL No.: MFCD00004624

Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 120…121 °C/13 mm
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004624
CAS Number: 16308-92-2
EINECS: 240-393-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR