Phenyltrimethylsilane 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L02876-06
L02876-14
CAAAAL02876-06EA
50.09
CAD
CAAAAL02876-06
CAAAAL02876-14
Phenyltrimethylsilane 99%
Phenyltrimethylsilane
Purity: 99%
CAS No.: 768-32-1
Molecular Formula: C9H14Si
Formula Weight: 150.30
Flash Point: 40°(104°F)
Refractive Index: 1.4900
Boiling Point: 169-170°
Beilstein Reference: 1855012
UN No.: 1993
MDL No.: MFCD00008271
Formula:
C₉H₁₄Si Boiling Pt: 169…170 °C Density: 0.871 Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008271 CAS Number: 768-32-1 EINECS: 212-192-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...