Ethyl methyl sulfide 97%

Supplier: Thermo Scientific Chemicals
Danger

L02859-14 L02859-22
CAAAAL02859-14EA 81.53 CAD
CAAAAL02859-14 CAAAAL02859-22
Ethyl methyl sulfide 97%
Ethyl methyl sulphide

Purity: 97% CAS No.: 624-89-5 Molecular Formula: C3H8S Formula Weight: 76.16 Flash Point: -15°(5°F) Refractive Index: 1.4392 Odor: Stench Melting Point: -106° Boiling Point: 66-67° Beilstein Reference: 1696871 UN No.: 1993 MDL No.: MFCD00009268

Formula: C₃H₈S
MW: 76.16 g/mol
Boiling Pt: 65
Melting Pt: -104.8
Density: 0.841
Flash Pt: –15 °C (5 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009268
CAS Number: 624-89-5
EINECS: 210-868-4
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR