5-Chloro-1-pentyne 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L02835-06
L02835-14
CAAAAL02835-06EA
78.37
CAD
CAAAAL02835-06
CAAAAL02835-14
5-Chloro-1-pentyne 98%
5-Chloro-1-pentyne
Purity: 98%
CAS No.: 14267-92-6
Molecular Formula: C5H7Cl
Formula Weight: 102.56
Storage Temperature: 2°C to 8°C
Flash Point: 15°(59°F)
Refractive Index: 1.4440
Odor: Recognizable
Boiling Point: 113°
Beilstein Reference: 1736710
UN No.: 1993
MDL No.: MFCD00001014
Formula:
C₅H₇Cl Boiling Pt: 113 °C Density: 0.968 Flash Pt: 15 °C (59 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001014 CAS Number: 14267-92-6 EINECS: 238-159-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...