1,3-Diphenylacetone 98+%

Supplier: Thermo Scientific Chemicals

L02830-14 L02830-22
CAAAAL02830-14EA 84.21 CAD
CAAAAL02830-14 CAAAAL02830-22
1,3-Diphenylacetone 98+%
1,3-Diphenylacetone

Purity: 98+% CAS No.: 102-04-5 Molecular Formula: C15H14O Formula Weight: 210.28 Flash Point: 110°(230°F) Melting Point: 32-36° Boiling Point: 330° Beilstein Reference: 974767 MDL No.: MFCD00004795

Formula: (C₆H₅)CH₂C(O)CH₂(C₆H₅)
Boiling Pt: 330 °C
Melting Pt: 32…36 °C
Density: 1.04
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004795
CAS Number: 102-04-5
EINECS: 203-000-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR