N-(Bromomethyl)phthalimide 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Bromomethyl)isoindoline-1,3-dione
L02818-09
L02818-18
CAAAAL02818-09EA
133.83
CAD
CAAAAL02818-09
CAAAAL02818-18
N-(Bromomethyl)phthalimide 95%
N-(Bromomethyl)phthalimide
Purity: 95%
CAS No.: 5332-26-3
Molecular Formula: C9H6BrNO2
Formula Weight: 240.07
Odor: Odorless
Melting Point: 144-150°
Beilstein Reference: 140943
MDL No.: MFCD00005897
Formula:
C₉H₆BrNO₂ MW: 240.06 g/mol Boiling Pt: 410 Melting Pt: 144…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005897 CAS Number: 5332-26-3 EINECS: 226-239-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...