(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2,2-Dimethyl-1,3-dioxolan-4-yl)methanol, 2,2-Dimethyl-1,3-dioxolan-4-ylmethanol, DL-1,2-Isopropylidenglycerol, 1,2-O-Isopropylideneglycerol, Solketal, (±)-1,2-O-Isopropylidenglycerol, DL-1,2-Isopropylideneglycerol

L02814-0E L02814-22 L02814-36
CAAAAL02814-0EEA 410.8 CAD
CAAAAL02814-0E CAAAAL02814-22 CAAAAL02814-36
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 97%
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol

Useful for synthesis of mono-, di- and triglycerides


Purity: 97% CAS No.: 100-79-8 Molecular Formula: C6H12O3 Formula Weight: 132.16 Flash Point: 80°(176°F) Refractive Index: 1.4340 Melting Point: -27° Boiling Point: 188-190° Beilstein Reference: 104465 Merck Reference: 14,5213 MDL No.: MFCD00063238

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 188
Melting Pt: –27 °C
Density: 1.064
Flash Pt: 80 °C (176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063238
CAS Number: 100-79-8
EINECS: 202-888-7
Merck Index: 14,5213

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR