Tetrabutylammonium hydroxide 40% (w/w) in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
L02809-0B
L02809-18
L02809-30
CAAAAL02809-0BEA
711
CAD
CAAAAL02809-0B
CAAAAL02809-18
CAAAAL02809-30
Tetrabutylammonium hydroxide 40% (w/w) in aqueous solution
Tetrabutylammonium hydroxide
CAS No.: 2052-49-5
Molecular Formula: C16H37NO
Formula Weight: 259.48
Flash Point: None
Refractive Index: 1.4080
Odor: Amine-like
pH: 14.0
Beilstein Reference: 3571228
UN No.: 3267
MDL No.: MFCD00009425
Formula:
C₁₆H₃₇NO Density: 0.990 Storage Temperature: Ambient |
MDL Number:
MFCD00009425 CAS Number: 2052-49-5 EINECS: 218-147-6 UN: 3267 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...