2-Iodo-2-methylpropane 95% stabilized, powder
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
tert-Butyl iodide, 2-Methyl-2-propyliodide
Stabilization:
Stabilised with copper
L02786-14
L02786-22
CAAAAL02786-14EA
125.14
CAD
CAAAAL02786-14
CAAAAL02786-22
2-Iodo-2-methylpropane 95% stabilized, powder
2-Iodo-2-methylpropane
Purity: 95%
CAS No.: 558-17-8
Molecular Formula: C4H9I
Formula Weight: 184.02
Flash Point: 7°(44°F)
Refractive Index: 1.4910
Odor: Recognizable
Melting Point: -38°
Boiling Point: 99-100°
Beilstein Reference: 505950
UN No.: 2391
MDL No.: MFCD00001068
Formula:
C₄H₉I MW: 184.02 g/mol Boiling Pt: 99…100 °C Melting Pt: –38 °C Density: 1.547 Flash Pt: 7 °C (44 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001068 CAS Number: 558-17-8 EINECS: 209-190-1 UN: 2391 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...