trans-4-(Trifluoromethyl)cinnamic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L02771-03
L02771-06
CAAAAL02771-03EA
59.57
CAD
CAAAAL02771-03
CAAAAL02771-06
trans-4-(Trifluoromethyl)cinnamic acid 98%
trans-4-(Trifluoromethyl)cinnamic acid
Purity: 98%
CAS No.: 16642-92-5
Molecular Formula: C10H7F3O2
Formula Weight: 216.16
Odor: Odorless
Melting Point: 230-234°
Beilstein Reference: 2452800
MDL No.: MFCD00002696
Formula:
C₁₀H₇F₃O₂ Melting Pt: 230…234 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002696 CAS Number: 16642-92-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...