4-Pentyn-2-ol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Pentyn-4-ol
L02752-06
L02752-14
CAAAAL02752-06EA
79.79
CAD
CAAAAL02752-06
CAAAAL02752-14
4-Pentyn-2-ol 97%
4-Pentyn-2-ol
Purity: 97%
CAS No.: 2117-11-5
Molecular Formula: C5H8O
Formula Weight: 84.12
Flash Point: 37°(98°F)
Refractive Index: 1.4380
Boiling Point: 126-127°
Beilstein Reference: 1734245
UN No.: 1987
MDL No.: MFCD00004555
Formula:
C₅H₈O Boiling Pt: 126…127 °C Density: 0.892 Flash Pt: 37 °C (98 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004555 CAS Number: 2117-11-5 EINECS: 218-321-1 UN: 1986 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...