Tris(pentafluorophenyl)phosphine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Perfluorotriphenylphosphine, Tris(perfluorophenyl)phosphine
L02748-03
L02748-06
CAAAAL02748-03EA
94.8
CAD
CAAAAL02748-03
CAAAAL02748-06
Tris(pentafluorophenyl)phosphine 97%
Tris(pentafluorophenyl)phosphine
Purity: 97%
CAS No.: 1259-35-4
Molecular Formula: C18F15P
Formula Weight: 532.15
Odor: Odorless
Melting Point: 115-119°
Beilstein Reference: 773243
MDL No.: MFCD00079654
Formula:
C₁₈F₁₅P MW: 532.15 g/mol Melting Pt: 115…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00079654 CAS Number: 1259-35-4 EINECS: 215-021-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...