Bis (8-hydroxyquinolinium) sulfate monohydrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L02743-14
L02743-22
CAAAAL02743-14EA
41.24
CAD
CAAAAL02743-14
CAAAAL02743-22
Bis (8-hydroxyquinolinium) sulfate monohydrate 99%
Bis (8-hydroxyquinolinium) sulphate monohydrate
Purity: 99%
CAS No.: 207386-91-2
Molecular Formula: C18H14N2O2.H2SO4.H2O
Formula Weight: 406.42 (388.40anhy)
Melting Point: 176-179°
Beilstein Reference: 3760550
Formula:
C₁₈H₁₆N₂O₆S·1H₂O Melting Pt: 176…179 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149610 CAS Number: 207386-91-2 EINECS: 205-137-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...